Anti-NET1 Goat Polyclonal Antibody

Supplier: Prosci
45-947
10112-018EA 701.94 USD
10112-018
Anti-NET1 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:32000. Western Blot: Approximately 85 kDa in human placenta and a very strong band at approximately 40-50 kDa in human skeletal muscle lysates after 0.1 ?g/mL staining.7 kDa according to NP_001040625.1 and of 61.9 kDa according to NP_005854.2. Immunohistochemistry: In paraffin embedded human tonsil shows staining of fibrous structures. Recommended concentration, 2.5 ?g/mL.

NET1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: NET1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR