Anti-NCF4 Goat Polyclonal Antibody

Supplier: Prosci
45-937
10116-384EA 701.94 USD
10116-384
Anti-NCF4 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:32000. Western Blot: Approximately 38 kDa band observed in lysates of Daudi lysate (calculated MW of 39.0 kDa according to NP_000622.2 and 38.8 kDa according to NP_038202.1). In transfected HEK293 transiently expressing P40PHOX a band of approximately 42 kDa is observed. This band is not observed in the non-transfected HEK293. Recommended concentration: 0.3-1 ?g/mL. Immunohistochemistry: In paraffin embedded human tonsil shows staining of the cytoplasm in some cells. Recommended concentration, 2-4 ?g/mL.

NCF4 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: NCF4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR