Anti-STXBP1 Goat Polyclonal Antibody

Supplier: Prosci
45-916
10116-358EA 701.94 USD
10116-358
Anti-STXBP1 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:16,000. Western Blot: Approximately 75 kDa band observed in human and mouse brain lysates (calculated MW of 68.7 kDa according to NP_003156.1). Recommended concentration: 0.1-0.3 ?g/mL. Immunohistochemistry: In paraffin embedded human brain cortex shows vesicular staining in neuronal bodies. Recommended concentration, 5-10 ?g/mL.

MUNC18 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: STXBP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR