Anti-MTMR9 Goat Polyclonal Antibody

Supplier: Prosci
45-912
10116-352EA 701.94 USD
10116-352
Anti-MTMR9 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:8000. Immunohistochemistry: Immunohistochemsitry: This antibody has been successfully used on rat Hypothalamus.

MTMR9 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: MTMR9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR