Anti-MERTK Goat Polyclonal Antibody

Supplier: Prosci
45-885
10116-306EA 701.94 USD
10116-306
Anti-MERTK Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:2000. Western Blot: Approximately 30 kDa band observed in human brain, human ovary, human kidney, MOLT-4 and U937 lysates at 1ug of primary. Please note that the band we observe is different from the predicted size of approximately 115 kDa according to NP_006334.

MERTK antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: MERTK
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR