Anti-IFIH1 Goat Polyclonal Antibody

Supplier: Prosci
45-874
10112-006EA 701.94 USD
10112-006
Anti-IFIH1 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:32000. Western Blot: No signal obtained yet but low background observed in human placenta lysate at up to 1 ?g/mL. Immunohistochemistry: In paraffin embedded human pancreas, shows strong cytoplasmic staining of some cells in the islets of Langerhans. Recommended concentration, 10 ?g/mL.

MDA5 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: IFIH1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR