Anti-MAML1 Goat Polyclonal Antibody

Supplier: Prosci
45-858
10116-264EA 701.94 USD
10116-264
Anti-MAML1 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:128,000. Western Blot: Approximately 110 kDa band observed in human skeletal muscle lysates (calculated MW of 108 kDa according to NP_055572.1). Recommended concentration: 0.1-0.3 ?g/mL. Immunohistochemistry: In paraffin embedded human placenta shows nuclear staining in trophoblasts. Recommended concentration, 2.5-3.8 ?g/mL.

MAML1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: MAML1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR