Anti-MAGOH Goat Polyclonal Antibody

Supplier: Prosci
45-856
10116-262EA 701.94 USD
10116-262
Anti-MAGOH Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:2000. Western Blot: Approximately 17 kDa band observed in nuclear HeLa lysate (calculated MW of 17.2 kDa according to NP_002361.1). Recommended concentration: 1-3 ?g/mL.

MAGOH antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: MAGOH
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR