Anti-M6PRBP1 Goat Polyclonal Antibody

Supplier: Prosci
45-852
10116-254EA 701.94 USD
10116-254
Anti-M6PRBP1 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:16,000. Western Blot: Approximately 48 kDa band observed in rat skin lysates (calculated MW of 47.3 kDa according to mouse NP_080112.1). Recommended concentration: 1-3 ?g/mL.

M6prbp1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: M6prbp1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR