Anti-LMO7 Goat Polyclonal Antibody

Supplier: Prosci
45-843
10116-238EA 701.94 USD
10116-238
Anti-LMO7 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:32,000. Western Blot: Approximately 70-75 kDa band and Approximately 30 kDa band observed in human lung lysates (predicted MWs of 92 kDa according to AAD33924.1 - however our result agrees with Putilina et al as below). A minor band of unknown identity was also consistently observed at approximately 50 kDa . Recommended for use at 0.5-2 ?g/mL.

LMO7 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: LMO7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR