Anti-LCN2 Goat Polyclonal Antibody

Supplier: Prosci
45-838
10111-998EA 701.94 USD
10111-998
Anti-LCN2 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:16000. Western Blot: Approximately 22 kDa band observed in human kidney lysates (calculated MW of 22.6 kDa according to NP_005555.2). Recommended concentration: 0.01-0.03 ?g/mL. Immunohistochemistry: In paraffin embedded human spleen shows staining of cytoplasm in mostly binucleated cells, and of nucleoli in some of the non-dividing cells. Recommended concentration, 2-3 ?g/mL.

Lipocalin 2 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: LCN2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR