Anti-LPIN3 Goat Polyclonal Antibody

Supplier: Prosci
45-837
10116-232EA 701.94 USD
10116-232
Anti-LPIN3 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:64000. Western Blot: Approximately 80 kDa and 60 kDa in human liver lysates after 0.1 ?g/mL antibody staining.6 kDa according to NP_075047.1. Both detected bands were successfully blocked by incubation with the immunizing peptide (and BLAST results with the immunizing peptide sequence did not identify any other proteins to explain the bands). Immunohistochemistry: In paraffin embedded human thyroid gland shows staining of cytoplasm and cell membrane of epithelial cells surrounding the follicles. Recommended concentration, 2-3 ?g/mL.

Lipin 3 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: LPIN3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR