Anti-LAMA3 Goat Polyclonal Antibody

Supplier: Prosci
45-819
10116-212EA 552.17 USD
10116-212
Anti-LAMA3 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:2,000. Western Blot: Preliminary experiments in human kidney, lung and skin lysates gave no specific signal but low background (at antibody concentration up to 1 ?g/mL).

LAMA3 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: LAMA3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR