Anti-KLF3 Goat Polyclonal Antibody

Supplier: Prosci
45-805
10116-190EA 701.94 USD
10116-190
Anti-KLF3 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:32,000. Western Blot: Approximately 40 kDa band observed in human heart lysates (predicted MW of 40 kDa according to NP_057615). Recommended for use at 1-3 ?g/mL. Please note that the 40 kDa Band is sometimes accompanied by a 80-90 kDa band of unknown identity.

KLF3 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: KLF3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR