Anti-KLF16 Goat Polyclonal Antibody

Supplier: Prosci
45-804
10116-188EA 701.94 USD
10116-188
Anti-KLF16 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:32000. Western Blot: Approximately 24 kDa band observed in human heart lysates (calculated MW of 25.4 kDa according to NP_114124.1). Recommended concentration: 0.3-1 ?g/mL.

KLF16 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: KLF16
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR