Anti-KALRN Goat Polyclonal Antibody

45-789
10116-164EA 625.43 USD
10116-164
Anti-KALRN Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:4,000. Western Blot: Approximately 200 kDa band observed in mouse brain lysates (calculated MW of 192 kDa according to XP_001001845). Recommended concentration: 1-3 ?g/mL.

Kalirin antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: KALRN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR