Anti-ITPR3 Goat Polyclonal Antibody

Supplier: Prosci
45-786
10116-158EA 552.02 USD
10116-158
Anti-ITPR3 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:128,000. Western Blot: Approximately 60 kDa band in human adipose, colon, duodenum, heart and liver lysates after 1 ?g/mL antibody staining.1. The 60 kDa band was successfully blocked by incubation with the immunizing peptide.

ITPR3 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: ITPR3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR