Anti-IRAK3 Goat Polyclonal Antibody

Supplier: Prosci
45-778
10116-144EA 701.94 USD
10116-144
Anti-IRAK3 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:64,000. Western Blot: Approximately 65 kDa band observed in rat liver lysates (calculated MW of 67.8 kDa according to NP_082955.2). Recommended concentration: 0.05-0.2 ?g/mL.

IRAK3 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: IRAK3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR