ELISA: Antibody detection limit dilution 1:32,000. Western Blot: Approximately 120 kDa band observed in human brain lysates (predicted MW of 119 kDa according to NP_006382.1). Recommended for use at 0.1-1 ?g/mL. Please note that an approximately 30 kDa band of unknown identity is also consistently observed.
Importin 7 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.
Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.
Type: Primary
Antigen: IPO7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human