Anti-IGFBP6 Goat Polyclonal Antibody

Supplier: Prosci
45-764
10116-120EA 701.94 USD
10116-120
Anti-IGFBP6 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:32,000. Western Blot: Approximately 25 kDa band observed in human adrenal gland lysates (calculated MW of 25.3 kDa according to NP_002169.1). In transfected HEK293 transiently expressing IGFBP6 a band of approximately 29 kDa is observed. This band is not observed in the non-transfected HEK293. Recommended concentration: 0.5-1.5 ?g/mL.

IGFBP6 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: IGFBP6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR