Anti-HOXB13 Goat Polyclonal Antibody

Supplier: Prosci
45-742
10116-086EA 701.94 USD
10116-086
Anti-HOXB13 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:128,000. Western Blot: Approximately 28 kDa band observed in lysates of cell lines A431 and HeLa (calculated MW of 30.7 kDa according to NP_006352.2). Recommended concentration: 0.1-0.3 ?g/mL.

Homeobox B13 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: HOXB13
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR