Anti-HRH1 Goat Polyclonal Antibody

Supplier: Prosci
45-732
10113-326EA 701.94 USD
10113-326
Anti-HRH1 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:16,000. Western Blot: Approximately 55 kDa band observed in human heart lysates (calculated MW of 55.8 kDa according to NP_000852.1). Recommended concentration: 0.5-1.5 ?g/mL.

Histamine Receptor H1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: HRH1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR