Anti-TAS1R2 Goat Polyclonal Antibody

Supplier: Prosci
45-695
10116-012EA 701.76 USD
10116-012
Anti-TAS1R2 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:32,000. Western Blot: Approximately 100 kDa band observed in human pancreas lysates (calculated MW of 95.2 kDa according to NP_689418.2). Recommended concentration: 1-3 ?g/mL.

GPR71 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: TAS1R2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR