Anti-GOLGA6B Goat Polyclonal Antibody

Supplier: Prosci
45-687
10116-002EA 701.94 USD
10116-002
Anti-GOLGA6B Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:1,000. Western Blot: Approximately 45 kDa band in human liver lysates after 1 ?g/mL antibody staining. The 45 kDa band was successfully blocked by incubation with the immunizing peptide.

GOLGA6 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: GOLGA6B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR