Anti-GJB2 Goat Polyclonal Antibody

Supplier: Prosci
45-673
10115-980EA 701.94 USD
10115-980
Anti-GJB2 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:128,000. Western Blot: Approximately 26 kDa band observed in mouse brain lysates (calculated MW of 26.4 kDa according to mouse NP_032151.1 and 26.2 kDa according to human NP_003995.2). Recommended concentration: 0.01-0.03 ?g/mL.

GJB2 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: GJB2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR