Anti-GHRL Goat Polyclonal Antibody

Supplier: Prosci
45-669
10115-974EA 701.94 USD
10115-974
Anti-GHRL Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:8000. Immunohistochemistry: In paraffin embedded human placenta shows staining of trophoblast cytoplasm. Recommended concentration, 2-5 ?g/mL.

Ghrelin Preproprotein antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: GHRL
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR