Anti-ARHGEF5 Goat Polyclonal Antibody

Supplier: Prosci
45-664
10115-966EA 701.94 USD
10115-966
Anti-ARHGEF5 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:16,000. Western Blot: Approximately 170 kDa band observed in human HeLa lysates (calculated MW of 177 kDa according to NP_005426.2). Recommended concentration: 0.1-0.3 ?g/mL.

GEF5 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: ARHGEF5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR