Anti-GALR2 Goat Polyclonal Antibody

Supplier: Prosci
45-654
10115-950EA 701.94 USD
10115-950
Anti-GALR2 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:4,000. Western Blot: Approximately 40 kDa band observed in human heart lysates (calculated MW of 41.7 kDa according to NP_003848.1). Recommended concentration: 0.3-1 ?g/mL.

GALR2 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: GALR2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR