Anti-GAD2 Goat Polyclonal Antibody

Supplier: Prosci
45-648
10115-942EA 701.94 USD
10115-942
Anti-GAD2 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:32,000. Western Blot: Approximately 65+70 kDa bands observed in human brain lysates. an additional band of 26 kDa is observed. All bands were blocked by the immunizing peptide (calculated MW of 65.4 kDa according to NP_000809.1). Recommended concentration: 0.1-0.3 ?g/mL.

GAD65 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: GAD2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR