Anti-FZD9 Goat Polyclonal Antibody

Supplier: Prosci
45-637
10112-700EA 701.94 USD
10112-700
Anti-FZD9 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:32,000. Immunohistochemistry: In paraffin embedded human testis shows acrosomal staining of spermatids. Recommended concentration, 5-10 ?g/mL.

Frizzled 9 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: FZD9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR