Anti-FPR2 Goat Polyclonal Antibody

45-629
10113-298EA 617.84 USD
10113-298
Anti-FPR2 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:2,000. Western Blot: Approximately 25 kDa band in human lymph node and tonsil lysates after 1 ?g/mL antibody staining.0 kDa according to NP_001453.1. The 25 kDa band was successfully blocked by incubation with the immunizing peptide.

FPRL1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: FPR2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR