Anti-FOXP4 Goat Polyclonal Antibody

Supplier: Prosci
45-627
10112-686EA 701.94 USD
10112-686
Anti-FOXP4 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:4,000. Western Blot: Band at approximately 75 kDa in lysates of FOXP4-transfected HEK293 (calculated MW of 73.5 kDa according to NP_001012426.1). Recommended concentration: 1-3 ?g/mL.

FOXP4 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: FOXP4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR