Anti-FOXP3 Goat Polyclonal Antibody

Supplier: Prosci
45-626
10113-296EA 701.94 USD
10113-296
Anti-FOXP3 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:8000. Western Blot: Approximately 45 kDa band observed in human and murine brain lysates (calculated MW of 47.3 kDa according to NP_473380.1). Recommended concentration: 1-3 ?g/mL. In paraffin embedded mouse thymus shows nuclei stained in select thymocytes, with some intensely stained areas. Recommended concentration, 4-6 ?g/mL.

FOXP3 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: Foxp3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR