Anti-FOXC1 Goat Polyclonal Antibody

Supplier: Prosci
45-603
10113-288EA 701.94 USD
10113-288
Anti-FOXC1 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:32,000. Western Blot: Approximately 55 kDa band observed in human bone marrow lysates (calculated MW of 56.8 kDa according to NP_001444.1). Recommended concentration: 0.5-1.5 ?g/mL. Immunohistochemistry: In paraffin embedded human kidney shows nuclear staining of some cells in the glomeruli. Recommended concentration, 3 ?g/mL.

FOXC1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: FOXC1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse, Zebrafish
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR