Anti-FHL1 Goat Polyclonal Antibody

Supplier: Prosci
45-591
10112-644EA 701.76 USD
10112-644
Anti-FHL1 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:32000. Western Blot: Approximately 30 kDa band observed in human muscle lysates (calculated MW of 31.9 kDa according to NP_001440). Recommended concentration: 0.05-0.3 ?g/mL. Immunohistochemistry: In paraffin embedded human testis shows fibrous staining in the cytoplasm of developing sperm cells. Recommended concentration, 2-2 ?g/mL.

FHL1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: FHL1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR