Anti-FGFR1 Goat Polyclonal Antibody

Supplier: Prosci

45-589
10112-640EA 560.36 USD
10112-640
Anti-FGFR1 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:32000. Western Blot: Approximately 75 kDa band observed in human breast lysates (calculated MW of 82.2 kDa according to NP_075593, 81.9 kDa according to NP_075594). Recommended concentration: 0.3-1 ?g/mL. Immunohistochemistry: In paraffin embedded human small intestine shows staining of the enterocytic cytoplasm. Recommended concentration, 4-6 ?g/mL.

FGFR1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: FGFR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR