Anti-F13A1 Goat Polyclonal Antibody

Supplier: Prosci

45-564
10112-602EA 560.36 USD
10112-602
Anti-F13A1 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:32,000. Western Blot: Approximately 90 kDa band observed in human placenta lysates (calculated MW of 83.3 kDa according to NP_000120.2). Recommended concentration: 0.05-0.2 ?g/mL.

Factor XIII alpH a antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: F13A1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR