Anti-FAF2 Goat Polyclonal Antibody

Supplier: Prosci

45-553
10112-582EA 560.36 USD
10112-582
Anti-FAF2 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:4,000. Western Blot: Approximately 50-55 kDa band observed in human testis, MOLT-4 and Jurkat lysates (predicted MW of 51 kDa according to NP_055428). Recommended for use at 1-3 ?g/mL.

ETEA antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: FAF2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR