Anti-ERG Goat Polyclonal Antibody

Supplier: Prosci
45-549
10112-576EA 701.94 USD
10112-576
Anti-ERG Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:16,000.Immunohistochemistry: In paraffin embedded human prostate shows nuclear staining in secretory cells of the glands. Recommended concentration, 4-8 ?g/mL.

ERG antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: ERG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR