Anti-EN2 Goat Polyclonal Antibody

Supplier: Prosci
45-537
10112-560EA 701.76 USD
10112-560
Anti-EN2 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:4000. Western Blot: Approximately 30 kDa band observed in mouse and rat brain lysates (calculated MW of 34.2 kDa according to human NP_001418.2 and of 33.8 kDa according to mouse NP_034264.1 and of 33.7 kDa according to rat NP_001102684.1). Recommended concentration: 0.1-0.3 ?g/mL.

Engrailed-2 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: EN2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR