Anti-ELMO2 Goat Polyclonal Antibody

Supplier: Prosci
45-531
10112-552EA 701.94 USD
10112-552
Anti-ELMO2 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:32000. Western Blot: Approximately 80 kDa band seen in human brain lysate. Recommended for use at 1-3 ?g/mL. Immunohistochemistry: In paraffin embedded human prostate shows pixulated staining in the cytoplasm of stroma cells. Recommended concentration, 3-6 ?g/mL.

ELMO2 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: ELMO2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR