Anti-ELMO1 Goat Polyclonal Antibody

Supplier: Prosci
45-530
10112-550EA 701.94 USD
10112-550
Anti-ELMO1 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:32000. Western Blot: Approximately 70 kDa and 50 kDa bands seen in Jurkat cell lysate. Recommended for use at 1-2 ?g/mL. Note the smaller isoform of this protein has a consensus glycosylation site, which may explain the higher than expected band size (50 kDa versus 30 kDa ). Glycosylation can affect migration of proteins on SDS-PAGE [Hames, B.D and Rickwood, D., eds. Gel electrophoresis of proteins: a practical approach. Oxford University Press].

ELMO1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: ELMO1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Cow, Zebrafish
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR