Anti-EGR2 Goat Polyclonal Antibody

Supplier: Prosci
45-521
10112-534EA 701.94 USD
10112-534
Anti-EGR2 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:64000. Western Blot: Approximately 55 kDa band observed in HEPG2 lysate (calculated MW of 50.3 kDa according to NP_000390.2). Recommended concentration: 0.03-0.1 ?g/mL. Immunohistochemistry: In paraffin embedded human colon shows nuclear staining of absorptive cells. Recommended concentration, 3-5 ?g/mL.

EGR2 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: EGR2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR