Anti-ECT2 Goat Polyclonal Antibody

Supplier: Prosci
45-517
10112-528EA 701.94 USD
10112-528
Anti-ECT2 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution >1:32,000. Western Blot: 100 kDa band reported in human U87-MG glioblastoma lysate at 2.5 ?g/mL. A 30 kDa band of unknown identity was also observed.

ECT2 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: ECT2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR