Anti-DUOX1 Goat Polyclonal Antibody

Supplier: Prosci

45-501
10112-498EA 560.36 USD
10112-498
Anti-DUOX1 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:32,000. Immunohistochemistry: In paraffin embedded human thyroid shows staining of cytoplasm in epithelial cells of the gland. Recommended concentration, 4-6 ?g/mL.

Dual oxidase 1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: DUOX1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR