Anti-DROSHA Goat Polyclonal Antibody

Supplier: Prosci
45-498
10115-932EA 701.94 USD
10115-932
Anti-DROSHA Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:32,000. Western Blot: Approximately 150 kDa band observed in human liver lysates (calculated MW of 151 kDa according to NP_081075.2 and 159 kDa according to human NP_037367.2). Recommended concentration: 0.5-1.5 ?g/mL.

Drosha antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: RNASEN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR