Anti-DRD5 Goat Polyclonal Antibody

Supplier: Prosci
45-497
10115-930EA 625.23 USD
10115-930
Anti-DRD5 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:128,000. Western Blot: Approximately 75-80 kDa band in lysates of human brain (hippocampus), kidney, spleen and thymus after 0.1 ?g/mL antibody staining.0 kDa according to NP_000789.1. The 75-80 kDa band was successfully blocked by incubation with the immunizing peptide.

DRD5 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: DRD5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR