Anti-DOK3 Goat Polyclonal Antibody

Supplier: Prosci
45-490
10115-918EA 701.94 USD
10115-918
Anti-DOK3 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution >1:32,000. Western Blot: Approximately 60-65 kDa band observed in human PBMC lysate (predicted MW of 57 kDa according to NP_079148.1). Recommended for use at 1-3 ?g/mL.

DOK3 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: DOK3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR