Anti-DOCK1 Goat Polyclonal Antibody

Supplier: Prosci
45-489
10115-916EA 701.94 USD
10115-916
Anti-DOCK1 Goat Polyclonal Antibody
Antibodies
Western Blot: Approximately 150-200 kDa band seen in A431 cell lysate. Recommended for use at 3-4 ?g/mL.

DOCK1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: DOCK1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR