Anti-DLC1 Goat Polyclonal Antibody

Supplier: Prosci
45-487
10115-914EA 701.76 USD
10115-914
Anti-DLC1 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:32,000. Western Blot: Approximately 28 kDa band in human liver and lung lysates as well as in different epithelial lung cell lines after 0.3 ?g/mL antibody staining.1 and 54.6 kDa according to NP_079043.2 (isotope 3). The 28 kDa band was successfully blocked by incubation with the immunizing peptide.

DLC1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: DLC1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR